Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM170A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TMEM170A - transmembrane protein 170A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304996.1 | 1677 | UTR 3 | NP_001291925.1 | |||
NM_001304997.1 | 1677 | UTR 3 | NP_001291926.1 | |||
NM_001304998.1 | 1677 | UTR 3 | NP_001291927.1 | |||
NM_145254.2 | 1677 | UTR 3 | NP_660297.1 | |||
XM_011522869.2 | 1677 | UTR 3 | XP_011521171.1 | |||
XM_017022939.1 | 1677 | UTR 3 | XP_016878428.1 | |||
XM_017022940.1 | 1677 | UTR 3 | XP_016878429.1 | |||
XM_017022941.1 | 1677 | UTR 3 | XP_016878430.1 | |||
XM_017022942.1 | 1677 | Intron | XP_016878431.1 | |||
XM_017022943.1 | 1677 | UTR 3 | XP_016878432.1 |