Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||
Literature Links: |
TMEM170A PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU) - Not Available | |||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TMEM170A - transmembrane protein 170A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304996.1 | 1100 | UTR 3 | NP_001291925.1 | |||
NM_001304997.1 | 1100 | UTR 3 | NP_001291926.1 | |||
NM_001304998.1 | 1100 | UTR 3 | NP_001291927.1 | |||
NM_145254.2 | 1100 | UTR 3 | NP_660297.1 | |||
XM_011522869.2 | 1100 | UTR 3 | XP_011521171.1 | |||
XM_017022939.1 | 1100 | UTR 3 | XP_016878428.1 | |||
XM_017022940.1 | 1100 | UTR 3 | XP_016878429.1 | |||
XM_017022941.1 | 1100 | UTR 3 | XP_016878430.1 | |||
XM_017022942.1 | 1100 | Intron | XP_016878431.1 | |||
XM_017022943.1 | 1100 | UTR 3 | XP_016878432.1 |
Set Membership: |
Validated |