Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604191 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF263 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF263 - zinc finger protein 263 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005741.4 | 12 | Intron | NP_005732.2 | |||
XM_006720831.1 | 12 | UTR 5 | XP_006720894.1 | |||
XM_006720832.1 | 12 | UTR 5 | XP_006720895.1 | |||
XM_006720833.1 | 12 | UTR 5 | XP_006720896.1 | |||
XM_006720834.3 | 12 | UTR 5 | XP_006720897.1 | |||
XM_011522344.1 | 12 | UTR 5 | XP_011520646.1 | |||
XM_017022850.1 | 12 | Intron | XP_016878339.1 |
Set Membership: |
HapMap |