Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTCTTAGAGGACACTATTCAAGC[C/T]ACTACACTTGGTTTCTTCATACACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603436 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF205 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF205 - zinc finger protein 205 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042428.1 | Intron | NP_001035893.1 | ||||
NM_001278158.1 | Intron | NP_001265087.1 | ||||
NM_003456.2 | Intron | NP_003447.2 | ||||
XM_005255558.2 | Intron | XP_005255615.1 |
ZNF205-AS1 - ZNF205 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |