Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611462 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFSD6L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MFSD6L - major facilitator superfamily domain containing 6 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIK3R6 - phosphoinositide-3-kinase regulatory subunit 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010855.3 | Intron | NP_001010855.1 | ||||
NM_001290211.1 | Intron | NP_001277140.1 | ||||
XM_011523677.2 | Intron | XP_011521979.1 | ||||
XM_011523678.2 | Intron | XP_011521980.1 | ||||
XM_011523680.2 | Intron | XP_011521982.1 | ||||
XM_011523681.2 | Intron | XP_011521983.1 | ||||
XM_011523683.2 | Intron | XP_011521985.1 | ||||
XM_011523686.2 | Intron | XP_011521988.1 | ||||
XM_017024228.1 | Intron | XP_016879717.1 | ||||
XM_017024229.1 | Intron | XP_016879718.1 | ||||
XM_017024230.1 | Intron | XP_016879719.1 | ||||
XM_017024231.1 | Intron | XP_016879720.1 |
Set Membership: |
HapMap |