Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612064 MIM: 139390 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GIGYF1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GIGYF1 - GRB10 interacting GYF protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022574.4 | 4686 | UTR 3 | NP_072096.2 | |||
XM_005250532.1 | 4686 | UTR 3 | XP_005250589.1 | |||
XM_011516472.1 | 4686 | UTR 3 | XP_011514774.1 | |||
XM_011516477.1 | 4686 | UTR 3 | XP_011514779.1 | |||
XM_017012526.1 | 4686 | UTR 3 | XP_016868015.1 | |||
XM_017012527.1 | 4686 | UTR 3 | XP_016868016.1 | |||
XM_017012528.1 | 4686 | UTR 3 | XP_016868017.1 | |||
XM_017012529.1 | 4686 | UTR 3 | XP_016868018.1 | |||
XM_017012530.1 | 4686 | UTR 3 | XP_016868019.1 | |||
XM_017012531.1 | 4686 | UTR 3 | XP_016868020.1 |
GNB2 - G protein subunit beta 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |