Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609252 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIPI PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LIPI - lipase I | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBM11 - RNA binding motif protein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320602.1 | Intron | NP_001307531.1 | ||||
NM_144770.4 | Intron | NP_658983.3 | ||||
XM_005260997.4 | Intron | XP_005261054.1 | ||||
XM_017028384.1 | Intron | XP_016883873.1 | ||||
XM_017028385.1 | Intron | XP_016883874.1 | ||||
XM_017028386.1 | Intron | XP_016883875.1 | ||||
XM_017028387.1 | Intron | XP_016883876.1 |
Set Membership: |
HapMap |