Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBM46 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RBM46 - RNA binding motif protein 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277171.1 | 23 | UTR 5 | NP_001264100.1 | |||
NM_001277173.1 | 23 | Intron | NP_001264102.1 | |||
NM_144979.4 | 23 | UTR 5 | NP_659416.1 | |||
XM_006714119.2 | 23 | UTR 5 | XP_006714182.1 | |||
XM_006714120.1 | 23 | UTR 5 | XP_006714183.1 | |||
XM_011531692.1 | 23 | Intron | XP_011529994.1 | |||
XM_011531693.1 | 23 | Intron | XP_011529995.1 | |||
XM_011531694.1 | 23 | Intron | XP_011529996.1 | |||
XM_011531695.2 | 23 | UTR 5 | XP_011529997.1 | |||
XM_011531696.1 | 23 | Intron | XP_011529998.1 | |||
XM_011531697.2 | 23 | UTR 5 | XP_011529999.1 |