Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGTGCAGTAGCCCTCTATGAAGA[C/T]TATGCTTAGGATAAGTATGAGAATG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300343 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAGEA10 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MAGEA10 - MAGE family member A10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011543.2 | 1133 | Missense Mutation | ATC,GTC | I,V 234 | NP_001011543.2 | |
NM_001251828.1 | 1133 | Missense Mutation | ATC,GTC | I,V 234 | NP_001238757.1 | |
NM_021048.4 | 1133 | Missense Mutation | ATC,GTC | I,V 234 | NP_066386.2 |
MAGEA10-MAGEA5 - MAGEA10-MAGEA5 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204811.2 | 1133 | Intron | NP_001191740.1 |
Set Membership: |
HapMap |