Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601529 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NR2C1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NR2C1 - nuclear receptor subfamily 2 group C member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032287.2 | Intron | NP_001027458.1 | ||||
NM_001127362.1 | Intron | NP_001120834.1 | ||||
NM_003297.3 | Intron | NP_003288.2 | ||||
XM_005269133.2 | Intron | XP_005269190.1 | ||||
XM_006719584.2 | Intron | XP_006719647.1 | ||||
XM_006719585.2 | Intron | XP_006719648.1 | ||||
XM_011538716.2 | Intron | XP_011537018.1 | ||||
XM_011538717.2 | Intron | XP_011537019.1 |
Set Membership: |
HapMap |