Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603832 MIM: 606862 MIM: 609519 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFA3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
NDUFA3 - NADH:ubiquinone oxidoreductase subunit A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004542.3 | Intron | NP_004533.1 | ||||
XM_017026833.1 | Intron | XP_016882322.1 |
OSCAR - osteoclast associated, immunoglobulin-like receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282349.1 | Intron | NP_001269278.1 | ||||
NM_001282350.1 | Intron | NP_001269279.1 | ||||
NM_130771.4 | Intron | NP_570127.3 | ||||
NM_133168.4 | Intron | NP_573398.2 | ||||
NM_133169.4 | Intron | NP_573399.2 | ||||
NM_206818.2 | Intron | NP_996554.2 |
TFPT - TCF3 fusion partner | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |