Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 426000 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDM5D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KDM5D - lysine demethylase 5D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146705.1 | 4233 | Silent Mutation | CCC,CCT | P,P 1287 | NP_001140177.1 | |
NM_001146706.1 | 4233 | Silent Mutation | CCC,CCT | P,P 1199 | NP_001140178.1 | |
NM_004653.4 | 4233 | Silent Mutation | CCC,CCT | P,P 1256 | NP_004644.2 | |
XM_005262560.2 | 4233 | Silent Mutation | CCC,CCT | P,P 1211 | XP_005262617.1 | |
XM_005262561.2 | 4233 | Silent Mutation | CCC,CCT | P,P 1179 | XP_005262618.1 | |
XM_011531468.2 | 4233 | Silent Mutation | CCC,CCT | P,P 1230 | XP_011529770.1 |