Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 426000 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDM5D PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KDM5D - lysine demethylase 5D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146705.1 | 5179 | UTR 3 | NP_001140177.1 | |||
NM_001146706.1 | 5179 | UTR 3 | NP_001140178.1 | |||
NM_004653.4 | 5179 | UTR 3 | NP_004644.2 | |||
XM_005262560.2 | 5179 | UTR 3 | XP_005262617.1 | |||
XM_005262561.2 | 5179 | UTR 3 | XP_005262618.1 | |||
XM_011531468.2 | 5179 | UTR 3 | XP_011529770.1 |
Set Membership: |
HapMap |