Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142703 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HRH2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HRH2 - histamine receptor H2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131055.1 | Intron | NP_001124527.1 | ||||
NM_022304.2 | Intron | NP_071640.1 | ||||
XM_005265904.3 | Intron | XP_005265961.1 | ||||
XM_006714864.3 | Intron | XP_006714927.1 | ||||
XM_006714865.3 | Intron | XP_006714928.1 | ||||
XM_011534548.2 | Intron | XP_011532850.1 | ||||
XM_011534549.2 | Intron | XP_011532851.1 | ||||
XM_017009426.1 | Intron | XP_016864915.1 |
Set Membership: |
HapMap |