Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610572 MIM: 603139 | ||||||||||||||||||||
Literature Links: |
MARVELD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MARVELD2 - MARVEL domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001038603.2 | 576 | Silent Mutation | GGA,GGC | G,G 138 | NP_001033692.2 | |
NM_001244734.1 | 576 | Silent Mutation | GGA,GGC | G,G 138 | NP_001231663.1 | |
XM_005248445.4 | 576 | Silent Mutation | GGA,GGC | G,G 138 | XP_005248502.1 | |
XM_005248446.4 | 576 | Silent Mutation | GGA,GGC | G,G 138 | XP_005248503.1 | |
XM_005248447.4 | 576 | Silent Mutation | GGA,GGC | G,G 138 | XP_005248504.1 |
RAD17 - RAD17 checkpoint clamp loader component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |