Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126391 MIM: 603602 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LIG1 - DNA ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000234.2 | 2904 | UTR 3 | NP_000225.1 | |||
NM_001289063.1 | 2904 | UTR 3 | NP_001275992.1 | |||
NM_001289064.1 | 2904 | UTR 3 | NP_001275993.1 | |||
NM_001320970.1 | 2904 | UTR 3 | NP_001307899.1 | |||
NM_001320971.1 | 2904 | UTR 3 | NP_001307900.1 | |||
XM_017026809.1 | 2904 | UTR 3 | XP_016882298.1 |
PLA2G4C - phospholipase A2 group IVC | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |