Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTAGAAGCCATTCTCAGACCCCA[A/G]TCACCCCCCACAAAGCCTTGATTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608197 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PGLYRP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PGLYRP3 - peptidoglycan recognition protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052891.2 | Intron | NP_443123.1 | ||||
XM_011509118.1 | Intron | XP_011507420.1 | ||||
XM_011509119.1 | Intron | XP_011507421.1 | ||||
XM_011509120.2 | Intron | XP_011507422.1 |