Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610018 MIM: 605272 MIM: 612485 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF40 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARHGEF40 - Rho guanine nucleotide exchange factor 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278529.1 | Intron | NP_001265458.1 | ||||
NM_001278530.1 | Intron | NP_001265459.1 | ||||
NM_018071.4 | Intron | NP_060541.3 | ||||
XM_005267844.2 | Intron | XP_005267901.1 | ||||
XM_011536937.2 | Intron | XP_011535239.1 | ||||
XM_011536938.2 | Intron | XP_011535240.1 | ||||
XM_017021434.1 | Intron | XP_016876923.1 | ||||
XM_017021435.1 | Intron | XP_016876924.1 | ||||
XM_017021436.1 | Intron | XP_016876925.1 | ||||
XM_017021437.1 | Intron | XP_016876926.1 |
RNASE8 - ribonuclease A family member 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |