Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610018 MIM: 605272 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF40 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ARHGEF40 - Rho guanine nucleotide exchange factor 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278529.1 | Intron | NP_001265458.1 | ||||
NM_001278530.1 | Intron | NP_001265459.1 | ||||
NM_018071.4 | Intron | NP_060541.3 | ||||
XM_005267844.2 | Intron | XP_005267901.1 | ||||
XM_011536937.2 | Intron | XP_011535239.1 | ||||
XM_011536938.2 | Intron | XP_011535240.1 | ||||
XM_017021434.1 | Intron | XP_016876923.1 | ||||
XM_017021435.1 | Intron | XP_016876924.1 | ||||
XM_017021436.1 | Intron | XP_016876925.1 | ||||
XM_017021437.1 | Intron | XP_016876926.1 |
NDRG2 - NDRG family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |