Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605036 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM253 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TMEM253 - transmembrane protein 253 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146683.1 | 560 | Silent Mutation | CTC,CTT | L,L 90 | NP_001140155.1 | |
XM_006720233.3 | 560 | Silent Mutation | CTC,CTT | L,L 90 | XP_006720296.1 | |
XM_011537078.2 | 560 | Silent Mutation | CTC,CTT | L,L 90 | XP_011535380.1 | |
XM_011537079.1 | 560 | Silent Mutation | CTC,CTT | L,L 90 | XP_011535381.1 | |
XM_011537081.2 | 560 | Silent Mutation | CTC,CTT | L,L 53 | XP_011535383.1 | |
XM_011537082.2 | 560 | Silent Mutation | CTC,CTT | L,L 53 | XP_011535384.1 | |
XM_011537083.2 | 560 | Silent Mutation | CTC,CTT | L,L 90 | XP_011535385.1 | |
XM_011537084.2 | 560 | Silent Mutation | CTC,CTT | L,L 90 | XP_011535386.1 | |
XM_011537085.2 | 560 | Silent Mutation | CTC,CTT | L,L 53 | XP_011535387.1 |
ZNF219 - zinc finger protein 219 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101672.1 | 560 | Intron | NP_001095142.1 | |||
NM_001102454.1 | 560 | Intron | NP_001095924.1 | |||
NM_016423.2 | 560 | Intron | NP_057507.2 | |||
XM_005267739.1 | 560 | Intron | XP_005267796.1 | |||
XM_006720163.2 | 560 | Intron | XP_006720226.1 | |||
XM_006720164.3 | 560 | Intron | XP_006720227.1 | |||
XM_011536825.1 | 560 | Intron | XP_011535127.1 | |||
XM_017021354.1 | 560 | Intron | XP_016876843.1 | |||
XM_017021355.1 | 560 | Intron | XP_016876844.1 |
Set Membership: |
HapMap |