Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600830 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIM26 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRIM26 - tripartite motif containing 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242783.1 | 3036 | UTR 3 | NP_001229712.1 | |||
NM_003449.4 | 3036 | UTR 3 | NP_003440.1 | |||
XM_005249374.2 | 3036 | UTR 3 | XP_005249431.1 | |||
XM_005249375.2 | 3036 | UTR 3 | XP_005249432.1 | |||
XM_005249376.2 | 3036 | UTR 3 | XP_005249433.1 | |||
XM_005249377.2 | 3036 | UTR 3 | XP_005249434.1 | |||
XM_005249378.2 | 3036 | UTR 3 | XP_005249435.1 | |||
XM_006715180.2 | 3036 | UTR 3 | XP_006715243.1 | |||
XM_011514859.1 | 3036 | UTR 3 | XP_011513161.1 | |||
XM_017011263.1 | 3036 | UTR 3 | XP_016866752.1 |
Set Membership: |
HapMap |