Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAGGTGTGTGTGTGTGTATGTAC[A/G]CAGTGTCTGTGTATTTAAAATCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600855 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DYRK1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DYRK1A - dual specificity tyrosine phosphorylation regulated kinase 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001396.3 | Intron | NP_001387.2 | ||||
NM_101395.2 | Intron | NP_567824.1 | ||||
NM_130436.2 | Intron | NP_569120.1 | ||||
NM_130438.2 | Intron | NP_569122.1 | ||||
XM_005260933.4 | Intron | XP_005260990.1 | ||||
XM_006723976.3 | Intron | XP_006724039.1 | ||||
XM_006723977.3 | Intron | XP_006724040.1 | ||||
XM_006723978.3 | Intron | XP_006724041.1 | ||||
XM_006723979.3 | Intron | XP_006724042.1 | ||||
XM_011529482.2 | Intron | XP_011527784.1 | ||||
XM_011529483.2 | Intron | XP_011527785.1 | ||||
XM_011529484.2 | Intron | XP_011527786.1 | ||||
XM_011529485.2 | Intron | XP_011527787.1 | ||||
XM_017028284.1 | Intron | XP_016883773.1 | ||||
XM_017028285.1 | Intron | XP_016883774.1 | ||||
XM_017028286.1 | Intron | XP_016883775.1 |