Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRMD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FRMD1 - FERM domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122841.2 | Intron | NP_001116313.1 | ||||
NM_024919.4 | Intron | NP_079195.3 | ||||
XM_011536137.1 | Intron | XP_011534439.1 | ||||
XM_011536138.1 | Intron | XP_011534440.1 | ||||
XM_011536140.1 | Intron | XP_011534442.1 | ||||
XM_011536141.1 | Intron | XP_011534443.1 | ||||
XM_011536142.2 | Intron | XP_011534444.1 | ||||
XM_011536143.1 | Intron | XP_011534445.1 | ||||
XM_011536145.1 | Intron | XP_011534447.1 | ||||
XM_011536146.2 | Intron | XP_011534448.1 | ||||
XM_017011317.1 | Intron | XP_016866806.1 | ||||
XM_017011318.1 | Intron | XP_016866807.1 |