Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTCATTCTCTAAACAACTAATC[A/G]GTTGTAGTGACTCTTGACTGGAACT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602667 MIM: 604598 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NBN PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NBN - nibrin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024688.2 | 3415 | UTR 3 | NP_001019859.1 | |||
NM_002485.4 | 3415 | UTR 3 | NP_002476.2 | |||
XM_011517045.1 | 3415 | UTR 3 | XP_011515347.1 | |||
XM_011517046.1 | 3415 | Intron | XP_011515348.1 | |||
XM_017013460.1 | 3415 | UTR 3 | XP_016868949.1 | |||
XM_017013461.1 | 3415 | UTR 3 | XP_016868950.1 | |||
XM_017013462.1 | 3415 | UTR 3 | XP_016868951.1 |
OSGIN2 - oxidative stress induced growth inhibitor family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126111.2 | 3415 | Intron | NP_001119583.1 | |||
NM_004337.2 | 3415 | Intron | NP_004328.1 |
Set Membership: |
HapMap |