Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601872 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC7A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC7A2 - solute carrier family 7 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008539.3 | Intron | NP_001008539.3 | ||||
NM_001164771.1 | Intron | NP_001158243.1 | ||||
NM_003046.5 | Intron | NP_003037.4 | ||||
XM_005273610.4 | Intron | XP_005273667.1 | ||||
XM_005273611.4 | Intron | XP_005273668.1 | ||||
XM_005273612.4 | Intron | XP_005273669.1 | ||||
XM_017013746.1 | Intron | XP_016869235.1 | ||||
XM_017013747.1 | Intron | XP_016869236.1 |
Set Membership: |
HapMap |