Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTGGCTCTCCTGGGCCTTGGGTC[A/T]ACCTTTTCCCCCAGTTGGTGAGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602442 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITSN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ITSN1 - intersectin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001132.1 | Intron | NP_001001132.1 | ||||
NM_003024.2 | Intron | NP_003015.2 | ||||
XM_005261026.2 | Intron | XP_005261083.1 | ||||
XM_005261027.1 | Intron | XP_005261084.1 | ||||
XM_005261031.1 | Intron | XP_005261088.1 | ||||
XM_011529691.1 | Intron | XP_011527993.1 | ||||
XM_011529692.1 | Intron | XP_011527994.1 | ||||
XM_011529693.2 | Intron | XP_011527995.1 | ||||
XM_017028428.1 | Intron | XP_016883917.1 | ||||
XM_017028429.1 | Intron | XP_016883918.1 | ||||
XM_017028430.1 | Intron | XP_016883919.1 | ||||
XM_017028431.1 | Intron | XP_016883920.1 | ||||
XM_017028432.1 | Intron | XP_016883921.1 | ||||
XM_017028433.1 | Intron | XP_016883922.1 | ||||
XM_017028434.1 | Intron | XP_016883923.1 | ||||
XM_017028435.1 | Intron | XP_016883924.1 | ||||
XM_017028436.1 | Intron | XP_016883925.1 | ||||
XM_017028437.1 | Intron | XP_016883926.1 | ||||
XM_017028438.1 | Intron | XP_016883927.1 | ||||
XM_017028439.1 | Intron | XP_016883928.1 | ||||
XM_017028440.1 | Intron | XP_016883929.1 | ||||
XM_017028441.1 | Intron | XP_016883930.1 |
Set Membership: |
HapMap |