Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604251 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CABIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CABIN1 - calcineurin binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199281.1 | Intron | NP_001186210.1 | ||||
NM_001201429.1 | Intron | NP_001188358.1 | ||||
NM_012295.3 | Intron | NP_036427.1 | ||||
XM_005261415.2 | Intron | XP_005261472.1 | ||||
XM_005261416.2 | Intron | XP_005261473.1 | ||||
XM_005261417.2 | Intron | XP_005261474.1 | ||||
XM_005261419.2 | Intron | XP_005261476.1 | ||||
XM_006724179.1 | Intron | XP_006724242.1 | ||||
XM_006724180.1 | Intron | XP_006724243.1 | ||||
XM_006724181.1 | Intron | XP_006724244.1 | ||||
XM_011530026.1 | Intron | XP_011528328.1 | ||||
XM_011530029.2 | Intron | XP_011528331.1 | ||||
XM_011530030.2 | Intron | XP_011528332.1 | ||||
XM_017028680.1 | Intron | XP_016884169.1 | ||||
XM_017028681.1 | Intron | XP_016884170.1 | ||||
XM_017028682.1 | Intron | XP_016884171.1 | ||||
XM_017028683.1 | Intron | XP_016884172.1 | ||||
XM_017028684.1 | Intron | XP_016884173.1 |
Set Membership: |
HapMap |