Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGACGTGTTATGTTCCACCTTGT[C/T]TGCCCCCAGCAAGGCCCTTGAATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102776 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADORA2A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADORA2A - adenosine A2a receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000675.5 | Intron | NP_000666.2 | ||||
NM_001278497.1 | Intron | NP_001265426.1 | ||||
NM_001278498.1 | Intron | NP_001265427.1 | ||||
NM_001278499.1 | Intron | NP_001265428.1 | ||||
NM_001278500.1 | Intron | NP_001265429.1 |
ADORA2A-AS1 - ADORA2A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPECC1L-ADORA2A - SPECC1L-ADORA2A readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |