Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607711 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIP2A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DIP2A - disco interacting protein 2 homolog A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146115.1 | Intron | NP_001139587.1 | ||||
NM_001146116.1 | Intron | NP_001139588.1 | ||||
NM_015151.3 | Intron | NP_055966.2 | ||||
NM_206889.2 | Intron | NP_996772.1 | ||||
NM_206890.2 | Intron | NP_996773.1 | ||||
NM_206891.2 | Intron | NP_996774.1 | ||||
XM_011529492.2 | Intron | XP_011527794.1 | ||||
XM_011529501.1 | Intron | XP_011527803.1 | ||||
XM_011529502.1 | Intron | XP_011527804.1 | ||||
XM_017028291.1 | Intron | XP_016883780.1 | ||||
XM_017028292.1 | Intron | XP_016883781.1 | ||||
XM_017028293.1 | Intron | XP_016883782.1 | ||||
XM_017028294.1 | Intron | XP_016883783.1 | ||||
XM_017028295.1 | Intron | XP_016883784.1 | ||||
XM_017028296.1 | Intron | XP_016883785.1 | ||||
XM_017028297.1 | Intron | XP_016883786.1 | ||||
XM_017028298.1 | Intron | XP_016883787.1 | ||||
XM_017028299.1 | Intron | XP_016883788.1 | ||||
XM_017028300.1 | Intron | XP_016883789.1 | ||||
XM_017028301.1 | Intron | XP_016883790.1 | ||||
XM_017028302.1 | Intron | XP_016883791.1 | ||||
XM_017028303.1 | Intron | XP_016883792.1 | ||||
XM_017028304.1 | Intron | XP_016883793.1 | ||||
XM_017028305.1 | Intron | XP_016883794.1 |
DIP2A-IT1 - DIP2A intronic transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |