Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 146770 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DRICH1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DRICH1 - aspartate rich 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016449.3 | Intron | NP_057533.2 | ||||
XM_011530206.1 | Intron | XP_011528508.1 | ||||
XM_011530207.2 | Intron | XP_011528509.1 | ||||
XM_011530208.2 | Intron | XP_011528510.1 | ||||
XM_011530209.2 | Intron | XP_011528511.1 | ||||
XM_011530210.2 | Intron | XP_011528512.1 | ||||
XM_017028813.1 | Intron | XP_016884302.1 |
IGLL1 - immunoglobulin lambda like polypeptide 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |