Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCACAGGAGTCAGAGCTCTGCA[C/T]GTCTGCACCATGTCCTGGGCTCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605141 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VPREB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
VPREB1 - pre-B lymphocyte 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303509.1 | 25 | Missense Mutation | ACG,ATG | T,M 1 | NP_001290438.1 | |
NM_007128.3 | 25 | UTR 5 | NP_009059.1 |
Set Membership: |
HapMap |