Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTTTTTCCCTTTTGGTGATGTT[A/G]AGGGTGTGGATTGAGGGGGTACATA
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 608592 | |||||||||||||||||||||||||||||
Literature Links: |
CTDSPL PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | ||||||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR - Not Available | ||||||||
AMR - Not Available |
CTDSPL - CTD small phosphatase like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008392.1 | Intron | NP_001008393.1 | ||||
NM_005808.2 | Intron | NP_005799.2 | ||||
XM_017005519.1 | Intron | XP_016861008.1 | ||||
XM_017005520.1 | Intron | XP_016861009.1 |