Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605310 MIM: 164177 MIM: 600912 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCHCR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCHCR1 - coiled-coil alpha-helical rod protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POU5F1 - POU class 5 homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173531.2 | 1054 | Intron | NP_001167002.1 | |||
NM_001285986.1 | 1054 | UTR 5 | NP_001272915.1 | |||
NM_001285987.1 | 1054 | Silent Mutation | CCC,CCG | P,P 17 | NP_001272916.1 | |
NM_002701.5 | 1054 | Intron | NP_002692.2 | |||
NM_203289.5 | 1054 | UTR 5 | NP_976034.4 |
PSORS1C3 - psoriasis susceptibility 1 candidate 3 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TCF19 - transcription factor 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |