Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGTCCCCTTCCAGGGAGGAGTGA[C/T]GAGAGGTGGTGGAAGCAGAGATTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606670 MIM: 607524 MIM: 607525 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PPP1R11 - protein phosphatase 1 regulatory inhibitor subunit 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF39 - ring finger protein 39 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025236.3 | Intron | NP_079512.2 | ||||
NM_170769.2 | Intron | NP_739575.2 | ||||
XM_017011325.1 | Intron | XP_016866814.1 | ||||
XM_017011326.1 | Intron | XP_016866815.1 |
ZNRD1 - zinc ribbon domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |