Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
28 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608692 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLZF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
BLZF1 - basic leucine zipper nuclear factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320972.1 | Intron | NP_001307901.1 | ||||
NM_001320973.1 | Intron | NP_001307902.1 | ||||
NM_003666.3 | Intron | NP_003657.1 | ||||
XM_005245561.4 | Intron | XP_005245618.1 | ||||
XM_011510092.2 | Intron | XP_011508394.1 | ||||
XM_017002645.1 | Intron | XP_016858134.1 |
CCDC181 - coiled-coil domain containing 181 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300968.1 | Intron | NP_001287897.1 | ||||
NM_001300969.1 | Intron | NP_001287898.1 | ||||
NM_021179.2 | Intron | NP_067002.1 | ||||
XM_005245383.1 | Intron | XP_005245440.1 | ||||
XM_011509827.2 | Intron | XP_011508129.1 | ||||
XM_017001938.1 | Intron | XP_016857427.1 | ||||
XM_017001939.1 | Intron | XP_016857428.1 | ||||
XM_017001940.1 | Intron | XP_016857429.1 |
Set Membership: |
HapMap Validated |