Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCACAGAATACGCGCTCCCTCCC[G/T]CCCCCTTCTCTGTCCCCCGCCTCTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614641 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAMP5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LAMP5 - lysosomal associated membrane protein family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199897.1 | 375 | UTR 5 | NP_001186826.1 | |||
NM_012261.3 | 375 | UTR 5 | NP_036393.1 |
LAMP5-AS1 - LAMP5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |