Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616727 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHF21B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PHF21B - PHD finger protein 21B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135862.2 | 2804 | UTR 3 | NP_001129334.1 | |||
NM_001242450.1 | 2804 | UTR 3 | NP_001229379.1 | |||
NM_001284296.1 | 2804 | UTR 3 | NP_001271225.1 | |||
NM_138415.4 | 2804 | UTR 3 | NP_612424.1 | |||
XM_006724122.1 | 2804 | UTR 3 | XP_006724185.1 | |||
XM_011529855.1 | 2804 | Intron | XP_011528157.1 |