Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGAAGAAAAGAAGACAAAGAAC[G/A]GGTAGGAAAATTAAAAAGGTAGGAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136132 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FMO3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
FMO3 - flavin containing monooxygenase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002294.2 | 87 | UTR 5 | NP_001002294.1 | |||
NM_001319173.1 | 87 | UTR 5 | NP_001306102.1 | |||
NM_001319174.1 | 87 | UTR 5 | NP_001306103.1 | |||
NM_006894.5 | 87 | UTR 5 | NP_008825.4 | |||
XM_011509345.2 | 87 | UTR 5 | XP_011507647.1 |
Set Membership: |
HapMap DME Validated Inventoried |