Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTTTCCTCGTTATCTGCCCGACA[A/G]TGGGGCGGGCATTCAGGGTCGTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609082 MIM: 603264 MIM: 607207 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXL16 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FBXL16 - F-box and leucine rich repeat protein 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
JMJD8 - jumonji domain containing 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105371184 - uncharacterized LOC105371184 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHBDL1 - rhomboid like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STUB1 - STIP1 homology and U-box containing protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR24 - WD repeat domain 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032259.3 | Intron | NP_115635.1 | ||||
XM_011522699.2 | Intron | XP_011521001.1 |