Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGACTGGGTGGAGGAATCTTCCC[C/G]TCACAATGTCCCCTGGCTAAAAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600423 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ECE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ECE1 - endothelin converting enzyme 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113347.1 | 4313 | UTR 3 | NP_001106818.1 | |||
NM_001113348.1 | 4313 | UTR 3 | NP_001106819.1 | |||
NM_001113349.1 | 4313 | UTR 3 | NP_001106820.1 | |||
NM_001397.2 | 4313 | UTR 3 | NP_001388.1 | |||
XM_006710398.2 | 4313 | UTR 3 | XP_006710461.1 | |||
XM_011540872.2 | 4313 | UTR 3 | XP_011539174.1 | |||
XM_011540873.2 | 4313 | UTR 3 | XP_011539175.1 | |||
XM_017000511.1 | 4313 | UTR 3 | XP_016856000.1 |