Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607995 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UNC93A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
UNC93A - unc-93 homolog A (C. elegans) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143947.1 | Intron | NP_001137419.1 | ||||
NM_018974.3 | Intron | NP_061847.2 | ||||
XM_011535905.2 | Intron | XP_011534207.1 | ||||
XM_011535906.2 | Intron | XP_011534208.1 | ||||
XM_011535907.2 | Intron | XP_011534209.1 | ||||
XM_011535908.2 | Intron | XP_011534210.1 | ||||
XM_017010958.1 | Intron | XP_016866447.1 |
Set Membership: |
HapMap Validated |