Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCGTCCGAGGCTCCCGCCCCCC[C/G]CCGAGCGAATGTGTCCCCGGAAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607477 MIM: 610230 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTSE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GTSE1 - G2 and S-phase expressed 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRMU - tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282782.1 | Intron | NP_001269711.1 | ||||
NM_001282783.1 | Intron | NP_001269712.1 | ||||
NM_001282784.1 | Intron | NP_001269713.1 | ||||
NM_001282785.1 | Intron | NP_001269714.1 | ||||
NM_018006.4 | Intron | NP_060476.2 | ||||
XM_005261678.1 | Intron | XP_005261735.1 | ||||
XM_005261681.1 | Intron | XP_005261738.1 | ||||
XM_011530271.2 | Intron | XP_011528573.1 | ||||
XM_011530272.1 | Intron | XP_011528574.1 | ||||
XM_011530273.1 | Intron | XP_011528575.1 | ||||
XM_011530274.2 | Intron | XP_011528576.1 | ||||
XM_011530275.1 | Intron | XP_011528577.1 |
Set Membership: |
HapMap |