Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGGCGAGTCTCTGGGGATTAGCA[A/G]GAGCCAACTGTTAAATATTTAGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612025 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IYD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IYD - iodotyrosine deiodinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164694.1 | Intron | NP_001158166.1 | ||||
NM_001164695.1 | Intron | NP_001158167.1 | ||||
NM_001318495.1 | Intron | NP_001305424.1 | ||||
NM_203395.2 | Intron | NP_981932.1 | ||||
XM_006715478.3 | Intron | XP_006715541.1 | ||||
XM_006715479.3 | Intron | XP_006715542.1 |
Set Membership: |
HapMap |