Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300545 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HS6ST2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HS6ST2 - heparan sulfate 6-O-sulfotransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077188.1 | 3826 | UTR 3 | NP_001070656.1 | |||
NM_147175.3 | 3826 | UTR 3 | NP_671704.3 | |||
XM_005262490.3 | 3826 | UTR 3 | XP_005262547.1 | |||
XM_005262491.2 | 3826 | UTR 3 | XP_005262548.1 | |||
XM_011531406.1 | 3826 | UTR 3 | XP_011529708.1 | |||
XM_011531407.2 | 3826 | Intron | XP_011529709.1 | |||
XM_011531408.2 | 3826 | Intron | XP_011529710.1 | |||
XM_017029944.1 | 3826 | UTR 3 | XP_016885433.1 | |||
XM_017029945.1 | 3826 | UTR 3 | XP_016885434.1 | |||
XM_017029946.1 | 3826 | UTR 3 | XP_016885435.1 |
Set Membership: |
HapMap |