Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613591 MIM: 613594 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTN2A2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
BTN2A2 - butyrophilin subfamily 2 member A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197237.1 | Intron | NP_001184166.1 | ||||
NM_001197238.1 | Intron | NP_001184167.1 | ||||
NM_001197239.1 | Intron | NP_001184168.1 | ||||
NM_001197240.1 | Intron | NP_001184169.1 | ||||
NM_006995.4 | Intron | NP_008926.2 | ||||
NM_181531.2 | Intron | NP_853509.1 | ||||
XM_005248797.3 | Intron | XP_005248854.1 | ||||
XM_005248798.4 | Intron | XP_005248855.1 | ||||
XM_006714953.3 | Intron | XP_006715016.1 | ||||
XM_006714955.3 | Intron | XP_006715018.1 | ||||
XM_011514228.2 | Intron | XP_011512530.1 | ||||
XM_011514229.2 | Intron | XP_011512531.1 | ||||
XM_011514231.2 | Intron | XP_011512533.1 | ||||
XM_017010166.1 | Intron | XP_016865655.1 | ||||
XM_017010167.1 | Intron | XP_016865656.1 | ||||
XM_017010168.1 | Intron | XP_016865657.1 | ||||
XM_017010169.1 | Intron | XP_016865658.1 |
Set Membership: |
HapMap Validated |