Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTCACTTAGATTGCTACTCAGA[A/G]AAAGGCAGGAAACAGTTTTGTTTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 172420 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLCG1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PLCG1 - phospholipase C gamma 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002660.2 | Intron | NP_002651.2 | ||||
NM_182811.1 | Intron | NP_877963.1 | ||||
XM_005260438.2 | Intron | XP_005260495.1 | ||||
XM_011528865.1 | Intron | XP_011527167.1 | ||||
XM_011528867.2 | Intron | XP_011527169.1 |
PLCG1-AS1 - PLCG1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |