Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601601 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TFAP2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TFAP2B - transcription factor AP-2 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003221.3 | Intron | NP_003212.2 | ||||
XM_011514837.2 | Intron | XP_011513139.1 | ||||
XM_017011233.1 | Intron | XP_016866722.1 | ||||
XM_017011234.1 | Intron | XP_016866723.1 | ||||
XM_017011235.1 | Intron | XP_016866724.1 |
Set Membership: |
Validated |