Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613004 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HTT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HTT - huntingtin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MSANTD1 - Myb/SANT DNA binding domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042690.1 | 1647 | Intron | NP_001036155.1 | |||
XM_011513465.2 | 1647 | UTR 5 | XP_011511767.1 | |||
XM_011513466.2 | 1647 | UTR 5 | XP_011511768.1 | |||
XM_011513467.2 | 1647 | UTR 5 | XP_011511769.1 | |||
XM_017008169.1 | 1647 | UTR 5 | XP_016863658.1 |