Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAACTATATACACATTCAGGAATA[C/T]ATAACAGCAAACTGTTTGGAGGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616474 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NKAPL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NKAPL - NFKB activating protein like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZSCAN26 - zinc finger and SCAN domain containing 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001023560.3 | Intron | NP_001018854.2 | ||||
NM_001111039.2 | Intron | NP_001104509.1 | ||||
NM_001287421.1 | Intron | NP_001274350.1 | ||||
NM_001287422.1 | Intron | NP_001274351.1 | ||||
NM_152736.5 | Intron | NP_689949.3 | ||||
XM_011514862.2 | Intron | XP_011513164.1 | ||||
XM_011514864.1 | Intron | XP_011513166.1 | ||||
XM_011514866.1 | Intron | XP_011513168.1 | ||||
XM_011514867.1 | Intron | XP_011513169.1 | ||||
XM_011514869.1 | Intron | XP_011513171.1 | ||||
XM_017011264.1 | Intron | XP_016866753.1 | ||||
XM_017011265.1 | Intron | XP_016866754.1 |