Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604896 MIM: 615958 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MKKS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MKKS - McKusick-Kaufman syndrome | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLX4IP - SLX4 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009608.2 | Intron | NP_001009608.1 | ||||
XM_017027647.1 | Intron | XP_016883136.1 | ||||
XM_017027648.1 | Intron | XP_016883137.1 | ||||
XM_017027649.1 | Intron | XP_016883138.1 | ||||
XM_017027650.1 | Intron | XP_016883139.1 | ||||
XM_017027651.1 | Intron | XP_016883140.1 | ||||
XM_017027652.1 | Intron | XP_016883141.1 | ||||
XM_017027653.1 | Intron | XP_016883142.1 |